Product Details

SNP ID
rs143451278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51888645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAGCTGGCATTGCACAGGAGA[C/T]GGCTGGACATGTCTGAGGCACTGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD33 PubMed Links

Gene Details

Gene
ANKRD33
Gene Name
ankyrin repeat domain 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130015.1 321 UTR 5 NP_001123487.1
NM_001304459.1 321 UTR 5 NP_001291388.1
NM_001304460.1 321 UTR 5 NP_001291389.1
NM_182608.3 321 Missense Mutation CGG,TGG R75W NP_872414.3
XM_017019258.1 321 Missense Mutation CGG,TGG R84W XP_016874747.1

View Full Product Details