Product Details

SNP ID
rs143456534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96205264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATCACCTCAGCACATCCCATTA[C/T]GAACATACAGAGAGGGGACCAAACC
Phenotype
MIM: 600247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELK3 PubMed Links

Gene Details

Gene
ELK3
Gene Name
ELK3, ETS transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303511.1 Intron NP_001290440.1
NM_005230.3 Intron NP_005221.2
XM_006719275.3 Intron XP_006719338.1

View Full Product Details