Product Details

SNP ID
rs143565495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10160370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGTGGGGCATCAAAGGAGAACC[A/G]TCCTCCCAGAGCCATGGGTAGCTGG
Phenotype
MIM: 602601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLR1 PubMed Links

Gene Details

Gene
OLR1
Gene Name
oxidized low density lipoprotein receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172632.1 631 Missense Mutation CGG,TGG R173W NP_001166103.1
NM_001172633.1 631 Intron NP_001166104.1
NM_002543.3 631 Silent Mutation GAC,GAT D219D NP_002534.1

View Full Product Details