Product Details

SNP ID
rs143830032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133121577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGTTTTAGGTTGAATCTTCTG[A/G]TCTGGACAGATGGCTTGGGGAATTC
Phenotype
MIM: 194538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF10 PubMed Links

Gene Details

Gene
ZNF10
Gene Name
zinc finger protein 10
There are no transcripts associated with this gene.

Gene
ZNF891
Gene Name
zinc finger protein 891
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277291.1 533 Silent Mutation GAC,GAT D114D NP_001264220.1
XM_017018666.1 533 Silent Mutation GAC,GAT D114D XP_016874155.1
XM_017018667.1 533 Silent Mutation GAC,GAT D114D XP_016874156.1
XM_017018668.1 533 Silent Mutation GAC,GAT D114D XP_016874157.1

View Full Product Details