Product Details

SNP ID
rs143901626
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122986553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGAGCCATAGGCCGCGTGCAC[G/T]CGCAGGTGCAGCTGTGGGGAGACTG
Phenotype
MIM: 607668 MIM: 608920
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ARL6IP4 PubMed Links
Additional Information
For this assay, SNP(s) [rs12811109] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ARL6IP4
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 4
There are no transcripts associated with this gene.

Gene
OGFOD2
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 2
There are no transcripts associated with this gene.

Gene
PITPNM2
Gene Name
phosphatidylinositol transfer protein membrane associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300801.1 4070 Silent Mutation NP_001287730.1
NM_020845.2 4070 Silent Mutation NP_065896.1
XM_011538590.2 4070 Silent Mutation XP_011536892.1

View Full Product Details