Product Details

SNP ID
rs144070931
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71125345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCATCCACAGATTCATTTGTTCC[C/T]GATCTGGCAATACAGGACCATAGAA
Phenotype
MIM: 600769
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TSPAN8 PubMed Links

Gene Details

Gene
TSPAN8
Gene Name
tetraspanin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004616.2 882 Missense Mutation AGG,GGG R235G NP_004607.1
XM_006719583.3 882 Missense Mutation AGG,GGG R235G XP_006719646.1
XM_017019913.1 882 Missense Mutation AGG,GGG R235G XP_016875402.1

View Full Product Details