Product Details

SNP ID
rs144086612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121416329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAGCAGCTACGGAAGGGCCCAA[C/G]ATAGTTTGTAAAGCCTGTGGGCTTT
Phenotype
MIM: 609078 MIM: 608299
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KDM2B PubMed Links

Gene Details

Gene
KDM2B
Gene Name
lysine demethylase 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005366.1 307 Intron NP_001005366.1
NM_032590.4 307 Intron NP_115979.3
XM_005253955.3 307 Intron XP_005254012.1
XM_005253956.3 307 Intron XP_005254013.1
XM_005253961.4 307 Intron XP_005254018.1
XM_011538867.2 307 Intron XP_011537169.1
XM_011538868.2 307 Intron XP_011537170.1
XM_011538869.2 307 Intron XP_011537171.1
XM_011538875.2 307 Intron XP_011537177.1
Gene
RNF34
Gene Name
ring finger protein 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256858.1 307 Intron NP_001243787.1
NM_025126.3 307 Missense Mutation AAC,AAG N59K NP_079402.2
NM_194271.2 307 Missense Mutation AAC,AAG N60K NP_919247.1

View Full Product Details