Product Details

SNP ID
rs144106525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109483701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTCTCTGTCGGAGTCGACTGCA[A/G]AGAGATATCAGGTAAGGGCTAGGAT
Phenotype
MIM: 613421 MIM: 608047
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD10 PubMed Links

Gene Details

Gene
KCTD10
Gene Name
potassium channel tetramerization domain containing 10
There are no transcripts associated with this gene.

Gene
UBE3B
Gene Name
ubiquitin protein ligase E3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270449.1 419 Silent Mutation CAA,CAG Q50Q NP_001257378.1
NM_001270450.1 419 Silent Mutation CAA,CAG Q50Q NP_001257379.1
NM_001270451.1 419 Silent Mutation CAA,CAG Q50Q NP_001257380.1
NM_130466.3 419 Silent Mutation CAA,CAG Q50Q NP_569733.2
NM_183415.2 419 Silent Mutation CAA,CAG Q50Q NP_904324.1
XM_005253987.2 419 Silent Mutation CAA,CAG Q50Q XP_005254044.1
XM_006719681.3 419 Silent Mutation CAA,CAG Q50Q XP_006719744.1
XM_006719682.2 419 Silent Mutation CAA,CAG Q50Q XP_006719745.1
XM_011538959.2 419 Silent Mutation CAA,CAG Q50Q XP_011537261.1
XM_011538961.1 419 Silent Mutation CAA,CAG Q50Q XP_011537263.1
XM_017020195.1 419 UTR 5 XP_016875684.1
XM_017020196.1 419 Silent Mutation CAA,CAG Q50Q XP_016875685.1

View Full Product Details