Product Details

SNP ID
rs144181122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55492981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCAGATGGTTGTAGCCTCTGCT[G/T]TATTAACCTTTATTATCACTCTTGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6C68 PubMed Links

Gene Details

Gene
OR6C68
Gene Name
olfactory receptor family 6 subfamily C member 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005519.2 604 Missense Mutation GTA,TTA V202L NP_001005519.2

View Full Product Details