Product Details

SNP ID
rs144242008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121311441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATTATATACTCATGAAACAGT[C/G]GATTCATCAAGATATAACAATTATA
Phenotype
MIM: 606948
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANAPC5 PubMed Links

Gene Details

Gene
ANAPC5
Gene Name
anaphase promoting complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137559.1 Intron NP_001131031.1
NM_016237.4 Intron NP_057321.2
XM_005253900.3 Intron XP_005253957.1
XM_017019423.1 Intron XP_016874912.1
XM_017019424.1 Intron XP_016874913.1

View Full Product Details