Product Details

SNP ID
rs144296189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:20369791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCGCTGCTGGCCGCCTGCTGCGG[C/G]GGGGAAGCGCTCGTCCAGATTGGGC
Phenotype
MIM: 123805
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDE3A PubMed Links

Gene Details

Gene
PDE3A
Gene Name
phosphodiesterase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000921.4 547 Silent Mutation GGC,GGG G169G NP_000912.3
NM_001244683.1 547 Intron NP_001231612.1
XM_017019420.1 547 Silent Mutation GGC,GGG G169G XP_016874909.1
XM_017019421.1 547 Intron XP_016874910.1

View Full Product Details