Product Details

SNP ID
rs144437095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48525731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCTCTTTGTGCTGTTCCTGGGG[A/G]TTTACCTCCTGACCATAATGGAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8S1 PubMed Links

Gene Details

Gene
OR8S1
Gene Name
olfactory receptor family 8 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005203.2 100 Missense Mutation ATT,GTT I34V NP_001005203.2

View Full Product Details