Product Details

SNP ID
rs144477274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109783665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTCCACTTGCGGGGGTAACCCT[A/G]CTGGTGGCCATCGCAGCGGGGGTTC
Phenotype
MIM: 605427
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRPV4 PubMed Links

Gene Details

Gene
TRPV4
Gene Name
transient receptor potential cation channel subfamily V member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177428.1 2470 Nonsense Mutation CAG,TAG Q811* NP_001170899.1
NM_001177431.1 2470 Nonsense Mutation CAG,TAG Q824* NP_001170902.1
NM_001177433.1 2470 Nonsense Mutation CAG,TAG Q751* NP_001170904.1
NM_021625.4 2470 Nonsense Mutation CAG,TAG Q858* NP_067638.3
NM_147204.2 2470 Nonsense Mutation CAG,TAG Q798* NP_671737.1
XM_005253918.1 2470 Nonsense Mutation CAG,TAG Q858* XP_005253975.1
XM_011538630.2 2470 Nonsense Mutation CAG,TAG Q909* XP_011536932.2
XM_011538631.2 2470 Nonsense Mutation CAG,TAG Q862* XP_011536933.2
XM_011538632.2 2470 Nonsense Mutation CAG,TAG Q849* XP_011536934.2
XM_011538633.2 2470 Nonsense Mutation CAG,TAG Q802* XP_011536935.2
XM_011538634.2 2470 Intron XP_011536936.2
XM_011538635.2 2470 Intron XP_011536937.1
XM_011538636.2 2470 Intron XP_011536938.1
XM_017019774.1 2470 Nonsense Mutation CAG,TAG Q858* XP_016875263.1

View Full Product Details