Product Details

SNP ID
rs144537165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132715019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGATGGCTCCCTGGAGAAGGACC[A/G]CACTCTGACCCCGCTGGGTATGTGG
Phenotype
MIM: 614939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGAM5 PubMed Links

Gene Details

Gene
PGAM5
Gene Name
PGAM family member 5, mitochondrial serine/threonine protein phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170543.1 423 Missense Mutation CAC,CGC H118R NP_001164014.1
NM_001170544.1 423 Missense Mutation CAC,CGC H118R NP_001164015.1
NM_138575.3 423 Missense Mutation CAC,CGC H118R NP_612642.2

View Full Product Details