Product Details

SNP ID
rs144618122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52544690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTGGCCACATAGCCACCGCTGA[A/C]CATGCTGGGCCGGAAGCCATAGACA
Phenotype
MIM: 608245
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRT71 PubMed Links

Gene Details

Gene
KRT71
Gene Name
keratin 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033448.2 1484 Missense Mutation GTC,TTC V472F NP_258259.1
XM_017018749.1 1484 Missense Mutation GTC,TTC V390F XP_016874238.1

View Full Product Details