Product Details

SNP ID
rs144629821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57091431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAGCATCCCGCTCTTCAAGAT[A/C]TCTGAGACTGCGGGTACCCGGAAAG
Phenotype
MIM: 602381 MIM: 616496 MIM: 601512
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NAB2 PubMed Links

Gene Details

Gene
NAB2
Gene Name
NGFI-A binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005967.3 768 Silent Mutation ATA,ATC I130I NP_005958.1
XM_005268894.2 768 Silent Mutation ATA,ATC I130I XP_005268951.1
Gene
NEMP1
Gene Name
nuclear envelope integral membrane protein 1
There are no transcripts associated with this gene.

Gene
STAT6
Gene Name
signal transducer and activator of transcription 6
There are no transcripts associated with this gene.

View Full Product Details