Product Details

SNP ID
rs144639569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2820773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCACTGGGTCTTCCTGAACTGA[C/T]GGTGTCTCAGCCAGGTTGTGCGTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITFG2 PubMed Links

Gene Details

Gene
ITFG2
Gene Name
integrin alpha FG-GAP repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018463.3 736 Missense Mutation ACG,ATG T199M NP_060933.3
XM_005253707.4 736 Missense Mutation ACG,ATG T199M XP_005253764.1
XM_005253709.4 736 Missense Mutation ACG,ATG T22M XP_005253766.1
XM_017019641.1 736 Intron XP_016875130.1
Gene
NRIP2
Gene Name
nuclear receptor interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details