Product Details

SNP ID
rs144688256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57225103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGCGCAGCGCACCAAGAGGAAG[A/C]CGTCCATCAAGGCGGCGCGCGCCAA
Phenotype
MIM: 604637 MIM: 138450
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NDUFA4L2 PubMed Links

Gene Details

Gene
NDUFA4L2
Gene Name
NDUFA4, mitochondrial complex associated like 2
There are no transcripts associated with this gene.

Gene
NXPH4
Gene Name
neurexophilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007224.3 458 Missense Mutation ACG,CCG T95P NP_009155.1
XM_017018747.1 458 Missense Mutation ACG,CCG T95P XP_016874236.1
Gene
SHMT2
Gene Name
serine hydroxymethyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details