Product Details

SNP ID
rs144785225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55637042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTGATCATCCTCCTCACACAG[A/G]TCAGCCCTGCCCTGCACTCCCCCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10P1 PubMed Links

Gene Details

Gene
OR10P1
Gene Name
olfactory receptor family 10 subfamily P member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206899.1 151 Missense Mutation ATC,GTC I51V NP_996782.1

View Full Product Details