Product Details

SNP ID
rs144951708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132773254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGAGACTCGTGCACCGTCAGGG[C/T]GTGCTCCTCCATCTGGCGCTGCAGG
Phenotype
MIM: 602581
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLGA3 PubMed Links

Gene Details

Gene
GOLGA3
Gene Name
golgin A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172557.1 2512 Intron NP_001166028.1
NM_005895.3 2512 Missense Mutation ACC,GCC T1450A NP_005886.2
XM_005266164.4 2512 Missense Mutation ACC,GCC T1410A XP_005266221.1
XM_005266167.4 2512 Missense Mutation ACC,GCC T1259A XP_005266224.1
XM_006719736.3 2512 Missense Mutation ACC,GCC T1450A XP_006719799.1
XM_006719737.3 2512 Missense Mutation ACC,GCC T1450A XP_006719800.1
XM_011534793.2 2512 Missense Mutation ACC,GCC T1359A XP_011533095.1
XM_011534794.2 2512 Missense Mutation ACC,GCC T802A XP_011533096.1

View Full Product Details