Product Details

SNP ID
rs144955552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:460460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCGCTTCCGGCTGCTGCTGGC[A/G]CTCGCCGTGGTGTCTGTGGGGCTCT
Phenotype
MIM: 612220
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GALNT3 PubMed Links

Gene Details

Gene
B4GALNT3
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173593.3 84 Silent Mutation GCA,GCG A28A NP_775864.3

View Full Product Details