Product Details

SNP ID
rs145077282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12661235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATTTGGTGGATTTGAGTTAATG[C/G]GCCAAGCAAGCTTTTTTTCCTTGGC
Phenotype
MIM: 602927
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPR19 PubMed Links

Gene Details

Gene
GPR19
Gene Name
G protein-coupled receptor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006143.2 3912 Missense Mutation CCC,CGC P405R NP_006134.1
XM_011520623.2 3912 Missense Mutation CCC,CGC P424R XP_011518925.1
XM_011520624.2 3912 Missense Mutation CCC,CGC P424R XP_011518926.1
XM_011520625.2 3912 Missense Mutation CCC,CGC P405R XP_011518927.1
XM_017019212.1 3912 Missense Mutation CCC,CGC P424R XP_016874701.1
XM_017019213.1 3912 Missense Mutation CCC,CGC P424R XP_016874702.1
XM_017019214.1 3912 Missense Mutation CCC,CGC P424R XP_016874703.1
XM_017019215.1 3912 Missense Mutation CCC,CGC P424R XP_016874704.1
XM_017019216.1 3912 Missense Mutation CCC,CGC P405R XP_016874705.1

View Full Product Details