Product Details

SNP ID
rs145100205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56319910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATCCAGACCCACCAGGTCCCCA[A/G]TCTGTGGCATCTCATTCAGCATCAG
Phenotype
MIM: 605861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPY2 PubMed Links

Gene Details

Gene
CNPY2
Gene Name
canopy FGF signaling regulator 2
There are no transcripts associated with this gene.

Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127460.2 3289 Missense Mutation NP_001120932.1
NM_001166279.1 3289 Missense Mutation NP_001159751.1
NM_014871.4 3289 Missense Mutation NP_055686.3

View Full Product Details