Product Details

SNP ID
rs145322170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:20369917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGTGCTGAGGCTGAGGCTGGG[C/T]GTCCTCATGATCGCCTTGACTAGCG
Phenotype
MIM: 123805
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE3A PubMed Links

Gene Details

Gene
PDE3A
Gene Name
phosphodiesterase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000921.4 673 Silent Mutation GGC,GGT G211G NP_000912.3
NM_001244683.1 673 Intron NP_001231612.1
XM_017019420.1 673 Silent Mutation GGC,GGT G211G XP_016874909.1
XM_017019421.1 673 Intron XP_016874910.1

View Full Product Details