Product Details

SNP ID
rs145468530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49838931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACTGGATCTATGTCGCAGCAGA[A/G]GAGACGGAATTCTCTTGAGGCATCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCDIN3D PubMed Links

Gene Details

Gene
BCDIN3D
Gene Name
BCDIN3 domain containing RNA methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181708.2 361 Missense Mutation CTC,TTC L107F NP_859059.1
Gene
BCDIN3D-AS1
Gene Name
BCDIN3D antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details