Product Details

SNP ID
rs145871604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7925916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGATGGGCTCTTGAACACCTGC[A/G]TCCTTGAAGATTCCTGTTGAGTAAT
Phenotype
MIM: 138170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC2A3 PubMed Links

Gene Details

Gene
SLC2A3
Gene Name
solute carrier family 2 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006931.2 Intron NP_008862.1

View Full Product Details