Product Details

SNP ID
rs146108048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121921606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGAAGAAACCACAGTAGAGCCC[C/T]AAGAAGTCACAGCGTCCATGATCCG
Phenotype
MIM: 603146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMD9 PubMed Links

Gene Details

Gene
PSMD9
Gene Name
proteasome 26S subunit, non-ATPase 9
There are no transcripts associated with this gene.

Gene
WDR66
Gene Name
WD repeat domain 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178003.1 473 Nonsense Mutation CAA,TAA Q101* NP_001171474.1
NM_144668.5 473 Nonsense Mutation CAA,TAA Q101* NP_653269.3

View Full Product Details