Product Details

SNP ID
rs146273754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109451042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGCTCTAGGCCCAAAGCCACAT[A/G]GCAGAAAGCACGTCTAGCAAAGCAG
Phenotype
MIM: 613421 MIM: 614636
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD10 PubMed Links

Gene Details

Gene
KCTD10
Gene Name
potassium channel tetramerization domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317395.1 1780 UTR 3 NP_001304324.1
NM_001317399.1 1780 UTR 3 NP_001304328.1
NM_031954.4 1780 UTR 3 NP_114160.1
Gene
MYO1H
Gene Name
myosin IH
There are no transcripts associated with this gene.

View Full Product Details