Product Details

SNP ID
rs146460124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9832369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTTACTGGGAATATCTGGAACC[A/G]TGAATGGTATTCTCACTTTGACTTT
Phenotype
MIM: 605029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLRF1 PubMed Links

Gene Details

Gene
KLRF1
Gene Name
killer cell lectin like receptor F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291822.1 381 Missense Mutation ATG,GTG M47V NP_001278751.1
NM_001291823.1 381 Missense Mutation ATG,GTG M47V NP_001278752.1
NM_016523.2 381 Missense Mutation ATG,GTG M47V NP_057607.1
XM_017019414.1 381 Missense Mutation ATG,GTG M47V XP_016874903.1
XM_017019415.1 381 Missense Mutation ATG,GTG M47V XP_016874904.1

View Full Product Details