Product Details

SNP ID
rs146463326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55400886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTGGCTGTCATGTCTTATGAT[C/T]GCTATGTGGCTATCTGCAAACCTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6C65 PubMed Links

Gene Details

Gene
OR6C65
Gene Name
olfactory receptor family 6 subfamily C member 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005518.1 358 Missense Mutation CGC,TGC R120C NP_001005518.1

View Full Product Details