Product Details

SNP ID
rs146534278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98613974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCGGAGCGTTGCTGTTCGATCAA[C/T]ACTGCTTGAAAGAAGATCACCTATA
Phenotype
MIM: 609861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IKBIP PubMed Links

Gene Details

Gene
IKBIP
Gene Name
IKBKB interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153687.3 896 Intron NP_710154.1
NM_201612.2 896 Missense Mutation ATT,GTT I222V NP_963906.1
NM_201613.2 896 UTR 3 NP_963907.1

View Full Product Details