Product Details

SNP ID
rs146578766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48525985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGCCTGCCTTCTCTCAGGGAT[A/G]GCCTATGACCGCCATGCTGCCATCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8S1 PubMed Links

Gene Details

Gene
OR8S1
Gene Name
olfactory receptor family 8 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005203.2 354 Missense Mutation ATA,ATG I118M NP_001005203.2

View Full Product Details