Product Details

SNP ID
rs146590886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:93408324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTAACATTTGTTACCAAGCAAT[C/G]CATTTATTTGGAAGGATACCAGTTA
Phenotype
MIM: 609229 MIM: 603679
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUDT4 PubMed Links

Gene Details

Gene
NUDT4
Gene Name
nudix hydrolase 4
There are no transcripts associated with this gene.

Gene
UBE2N
Gene Name
ubiquitin conjugating enzyme E2 N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003348.3 2540 UTR 3 NP_003339.1
XM_011538719.2 2540 Intron XP_011537021.1
XM_017019918.1 2540 Intron XP_016875407.1

View Full Product Details