Product Details

SNP ID
rs146768121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10159988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATATATGCACAGGTACCTGAAGG[A/G]TATGTCTGGGAGACAGCGCCTCGGA
Phenotype
MIM: 602601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLR1 PubMed Links

Gene Details

Gene
OLR1
Gene Name
oxidized low density lipoprotein receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172632.1 688 UTR 3 NP_001166103.1
NM_001172633.1 688 UTR 3 NP_001166104.1
NM_002543.3 688 Silent Mutation TAC,TAT Y238Y NP_002534.1

View Full Product Details