Product Details

SNP ID
rs146794656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14787363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTTGTATCATCCGCCTTGGGT[C/T]GCTGAATCAAGTTGGGTGGGGCACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf60 PubMed Links

Gene Details

Gene
C12orf60
Gene Name
chromosome 12 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175874.3 Intron NP_787070.2
XM_005253322.4 Intron XP_005253379.1
XM_011520568.2 Intron XP_011518870.1
XM_011520569.2 Intron XP_011518871.1
XM_017018871.1 Intron XP_016874360.1
XM_017018872.1 Intron XP_016874361.1
XM_017018873.1 Intron XP_016874362.1
XM_017018874.1 Intron XP_016874363.1
Gene
H2AFJ
Gene Name
H2A histone family member J
There are no transcripts associated with this gene.

Gene
WBP11
Gene Name
WW domain binding protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016312.2 Intron NP_057396.1

View Full Product Details