Product Details

SNP ID
rs146824657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:80708579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCCAGTGGCCAAGTGTTTCCG[A/C]TCATTCCAGGGGGCTCGTGATAACG
Phenotype
MIM: 159990 MIM: 159991
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYF5 PubMed Links

Gene Details

Gene
MYF5
Gene Name
myogenic factor 5
There are no transcripts associated with this gene.

Gene
MYF6
Gene Name
myogenic factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002469.2 666 Missense Mutation GAT,GCT D192A NP_002460.1

View Full Product Details