Product Details

SNP ID
rs147047550
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55332212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGATTACTGTGCTTCCAACGTCA[C/T]TGATCACTTTGCATGTGACTATTTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR6C3 PubMed Links

Gene Details

Gene
OR6C3
Gene Name
olfactory receptor family 6 subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054104.1 512 Missense Mutation ACT,ATT T171I NP_473445.1

View Full Product Details