Product Details

SNP ID
rs147053469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49963633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCCCTGTCCATTGGCCTGTCT[A/G]TCACCCTGGGCCACCTTGTCGGAGT
Phenotype
MIM: 107777 MIM: 600442 MIM: 601383
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AQP2 PubMed Links

Gene Details

Gene
AQP2
Gene Name
aquaporin 2
There are no transcripts associated with this gene.

Gene
AQP5
Gene Name
aquaporin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001651.3 1027 Missense Mutation ATC,GTC I169V NP_001642.1
XM_005268838.2 1027 Missense Mutation ATC,GTC I169V XP_005268895.1
Gene
AQP6
Gene Name
aquaporin 6
There are no transcripts associated with this gene.

Gene
LOC101927318
Gene Name
uncharacterized LOC101927318
There are no transcripts associated with this gene.

View Full Product Details