Product Details

SNP ID
rs147328826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:65170466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGCATCGGCCCAGACGAACCCA[C/T]AGTAAGCCTCTCCCCCCGCTGACTG
Phenotype
MIM: 607844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LEMD3 PubMed Links

Gene Details

Gene
LEMD3
Gene Name
LEM domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167614.1 896 Silent Mutation CAC,CAT H290H NP_001161086.1
NM_014319.4 896 Silent Mutation CAC,CAT H290H NP_055134.2

View Full Product Details