Product Details

SNP ID
rs147343644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14771070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTACCCAGCCCCTTGCCACCTTT[A/G]CCTCGCCCAGACATTCTGAAATCAC
Phenotype
MIM: 615069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf60 PubMed Links

Gene Details

Gene
C12orf60
Gene Name
chromosome 12 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175874.3 62 Intron NP_787070.2
XM_005253322.4 62 Intron XP_005253379.1
XM_011520568.2 62 Intron XP_011518870.1
XM_011520569.2 62 Intron XP_011518871.1
XM_017018871.1 62 Intron XP_016874360.1
XM_017018872.1 62 Intron XP_016874361.1
XM_017018873.1 62 Intron XP_016874362.1
XM_017018874.1 62 Intron XP_016874363.1
Gene
H2AFJ
Gene Name
H2A histone family member J
There are no transcripts associated with this gene.

Gene
HIST4H4
Gene Name
histone cluster 4, H4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175054.2 62 Silent Mutation GGC,GGT G5G NP_778224.1
Gene
LOC105369669
Gene Name
uncharacterized LOC105369669
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011520896.2 62 Intron XP_011519198.1

View Full Product Details