Product Details

SNP ID
rs147364129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8058840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCCTCCAGAATTCCCTGAATG[A/G]ACTGCCTTGCTTTCTTCCTAAAATC
Phenotype
MIM: 605246
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3AR1 PubMed Links

Gene Details

Gene
C3AR1
Gene Name
complement component 3a receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001326475.1 1550 Missense Mutation TCC,TTC S449F NP_001313404.1
NM_001326477.1 1550 Missense Mutation TCC,TTC S449F NP_001313406.1
NM_004054.3 1550 Missense Mutation TCC,TTC S449F NP_004045.1
Gene
FOXJ2
Gene Name
forkhead box J2
There are no transcripts associated with this gene.

View Full Product Details