Product Details

SNP ID
rs147640045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2799092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCCTCTCTTTCATGCCAGTTGC[A/C]CTGGAAGGGTACTACAAGGACAAGC
Phenotype
MIM: 600611
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FKBP4 PubMed Links

Gene Details

Gene
FKBP4
Gene Name
FK506 binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002014.3 717 Silent Mutation GCA,GCC A173A NP_002005.1
XM_011520929.2 717 Silent Mutation GCA,GCC A128A XP_011519231.1

View Full Product Details