Product Details

SNP ID
rs147643341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111644433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTTTTTGGTCACAGGTCTCCTT[C/T]AGCACCCTCTCCTCCTCTTTTAGCT
Phenotype
MIM: 604986
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRAP PubMed Links

Gene Details

Gene
BRAP
Gene Name
BRCA1 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006768.4 1540 Silent Mutation CTA,CTG L515L NP_006759.3
XM_005253944.4 1540 Silent Mutation CTA,CTG L556L XP_005254001.1
XM_011538789.2 1540 Silent Mutation CTA,CTG L228L XP_011537091.1
XM_017019992.1 1540 Silent Mutation CTA,CTG L461L XP_016875481.1
XM_017019993.1 1540 Silent Mutation CTA,CTG L366L XP_016875482.1
XM_017019994.1 1540 Silent Mutation CTA,CTG L366L XP_016875483.1
XM_017019995.1 1540 Silent Mutation CTA,CTG L366L XP_016875484.1

View Full Product Details