Product Details

SNP ID
rs147676166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:66123947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTGCCTTTGGTTCATCCATA[C/T]TGGGTACTGTCCATGCTGGTCTAGA
Phenotype
MIM: 616998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LLPH PubMed Links

Gene Details

Gene
LLPH
Gene Name
LLP homolog, long-term synaptic facilitation
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032338.3 Intron NP_115714.1
Gene
LLPH-AS1
Gene Name
LLPH antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details