Product Details
- SNP ID
-
rs147835463
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52544598 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTCCCTAGGGTGTCTTTGTAATC[A/G]TTGGCACTGCCCCGGCTCCTGCCCT
- Phenotype
-
MIM: 608245
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT71
PubMed Links
Gene Details
- Gene
- KRT71
- Gene Name
- keratin 71
View Full Product Details