Product Details

SNP ID
rs148000749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48569958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGACAGCTCACATTTTGTGAA[C/T]TGCTTGGCCAGGATGGCAGGGAACA
Phenotype
MIM: 149750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LALBA PubMed Links

Gene Details

Gene
LALBA
Gene Name
lactalbumin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002289.2 167 Silent Mutation CAA,CAG Q21Q NP_002280.1
XM_006719395.3 167 Silent Mutation CAA,CAG Q28Q XP_006719458.1

View Full Product Details