Product Details

SNP ID
rs148049721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52608296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGCCTGGTCCTGGCTTCCCCA[C/T]GGGGGCTACAGTTCCCACTGCCAGT
Phenotype
MIM: 608246 MIM: 608247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT72 PubMed Links

Gene Details

Gene
KRT72
Gene Name
keratin 72
There are no transcripts associated with this gene.

Gene
KRT73
Gene Name
keratin 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175068.2 3036 Missense Mutation CAT,CGT H508R NP_778238.1
XM_011538257.2 3036 Missense Mutation CAT,CGT H476R XP_011536559.1
Gene
KRT73-AS1
Gene Name
KRT73 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details