Product Details

SNP ID
rs148094880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54499424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCATGGAACCATCTCTCAAGTAT[A/G]TCAACAAGAAATTTCCCAACATAGA
Phenotype
MIM: 141180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCKAP1L PubMed Links

Gene Details

Gene
NCKAP1L
Gene Name
NCK associated protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184976.1 122 Missense Mutation ATC,GTC I8V NP_001171905.1
NM_005337.4 122 Missense Mutation ATC,GTC I58V NP_005328.2

View Full Product Details