Product Details

SNP ID
rs148097034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132714886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACCACTGTCTCTGATCAACGTG[C/T]GGAAGAGGAACGTGGAATCTGGGGA
Phenotype
MIM: 614939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGAM5 PubMed Links

Gene Details

Gene
PGAM5
Gene Name
PGAM family member 5, mitochondrial serine/threonine protein phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170543.1 290 Missense Mutation CGG,TGG R74W NP_001164014.1
NM_001170544.1 290 Missense Mutation CGG,TGG R74W NP_001164015.1
NM_138575.3 290 Missense Mutation CGG,TGG R74W NP_612642.2
Gene
PXMP2
Gene Name
peroxisomal membrane protein 2
There are no transcripts associated with this gene.

View Full Product Details