Product Details

SNP ID
rs148452184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:85283701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAGAAGGGAGAGCTGGATGAAC[G/T]TGGGGATAAATGTGATAGCAATGTA
Phenotype
MIM: 601527
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALX1 PubMed Links

Gene Details

Gene
ALX1
Gene Name
ALX homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006982.2 385 Missense Mutation CGT,CTT R119L NP_008913.2
XM_011538782.2 385 Missense Mutation CGT,CTT R24L XP_011537084.1

View Full Product Details